ContactUsLinkComponent
Standards for 2100N and 2100N Is
  MSPP-2897200
 :  
undefined
Standards for 2100N and 2100N Is
  MSPP-2897200
 :  2897200
 :  
  • Turbidity:
    0.1 NTU
    3 NTU
    6 NTU
    18 NTU
    30 NTU
  • Description:
    Stablcal® Turbidity Standards Calibration Kit
  • Cat. No.:
    MSPP-2897200