PCR Plates with Full Skirt - Low Profile, 384-Well
New Product
SARSTEDT PCR consumables are compatible with standard thermocyclers. Uniform and extremely thin well walls facilitate consistently homogeneous and rapid heat transfer. During production, each individual well is tested to ensure it is 100% leak-proof.
- Volume: 40 µl
- 384-well, low profile design
- Polypropylene material
- Optimized white color variant for maximum detection sensitivity in qPCR
- For low-volume reactions and fast PCR
- PCR plate 72. 1985.202 compatible with Roche LightCycler® 480
- Constant well and plate uniformity minimizes data variability
SARSTEDT PCR plates are 'PCR Performance Tested' certified and guaranteed to be DNA-free, free from DNase/RNase, and free from PCR inhibitors.