TNTplus™ Nitrogen Tests
Product Details & Documents
These 'Test N Tube' are used for the determination of nitrogen by the persulfate digestion method.
- Test 'N Tube products provide safe, convenient testing
- TNT plus vials reduce errors with bar coded vials and RFID
- TNT plus vials require no reagent blank
Specifications
Specifications
- Chemical method:Persulfate digestion
- Description:Nitrogen TNT plus kit, total, UHR
- Range:20 - 100 mg/L