Orders LinkContactUsLinkComponent
COD2 Digestion Vials
COD2 Digestion Vials
COD2 digestion vials, UHR, mercury-free, Reactor digestion, 200 - 15000 mg/L
Catalog # MSPP-2834325
Marketsource
Supplier:   HACH COMPANY MS
Supplier #:  2834325
COD2 Digestion Vials
Catalog # MSPP-2834325
Supplier:  HACH COMPANY MS
Marketsource
CAS Number:  
COD2 Digestion Vials
COD2 Digestion Vials
COD2 digestion vials, UHR, mercury-free, Reactor digestion, 200 - 15000 mg/L
CAS Number:  
Price
$101.34
Log in for availability
Each Pack of 25
Minimum order quantity is 1
Product Details & Documents
These vials are ideal for ultra-high range, mercury-free chemical oxygen demand determinations.
Specifications
Specifications
  • Chemical method:
    Reactor digestion
  • Description:
    COD2 digestion vials, UHR, mercury-free
  • Range:
    200 - 15000 mg/L
Specifications