Transfer Pipettes, with Graduations
Supplier: SARSTEDT INC
New Product
Certificates
About this item
Dosing, transferring, and measuring liquids are routine procedures in almost every laboratory. SARSTEDT offers a wide range of products for every application, from high-quality pipette tips and serological pipettes to transfer pipettes, for reliable and reproducible results.
- CE-certified in vitro diagnostic device, suitable for transferring liquid diagnostic sample material
- 0.5 ml graduations from 0.5 to 3.0 ml
- Made of polyethylene without any environmentally harmful substances added
- No risk of injury or infection as it is unbreakable
- Suction bulb can be easily squeezed to ensure effortless pipetting
SARSTEDT transfer pipettes are essential for the safe and practical transfer of liquids in laboratories. The right pipette type for a variety of applications can be found in this standard product range.