Orders LinkContactUsLinkComponent
Low Binding Reaction Tubes
Supplier:   SARSTEDT INC
Low Binding Reaction Tubes
Supplier:  SARSTEDT INC
Certificates
Low Binding Reaction Tubes
Low Binding Reaction Tubes
6 Options Available Below
Product Details & Documents
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • SafeSeal polypropylene caps attached
  • Reduced binding properties for higher recovery rates
  • PCR Performance Tested certified
  • Secure seal even under thermal and mechanical stress
  • High centrifugation up to at least 20,000 ×g
This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base, with or without skirted base, with push cap or screw cap, with or without print or labels.
Product Family Options
Product Information
  • Color
    Volume
    Cap type
    Sterility
    Availability
    Price
  • 77588-908
    Low protein-binding surface
    Low protein-binding surface
    Transparent
    0.5 ml
    Transparent, PP
    Non sterile (PCR performance tested)
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 300
    Pack of 300 - $55.16
    $55.16
    Log in for availability
    Case of 1200
    Case of 1200 - $220.64
    $220.64
    Pack of 300
       $55.16
    Case of 1200
       $220.64
    Supplier Number: 72.704.600
  • 77588-910
    Low DNA-binding surface
    Low DNA-binding surface
    Transparent
    0.5 ml
    Transparent, PP
    Non sterile (PCR performance tested)
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 300
    Pack of 300 - $50.24
    $50.24
    Log in for availability
    Case of 1200
    Case of 1200 - $200.96
    $200.96
    Pack of 300
       $50.24
    Case of 1200
       $200.96
    Supplier Number: 72.704.700
  • 77588-930
    Low protein-binding surface
    Low protein-binding surface
    Transparent
    1.5 ml
    Transparent, PP
    Non sterile (PCR performance tested)
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 200
    Pack of 200 - $33.50
    $33.50
    Log in for availability
    Case of 800
    Case of 800 - $134.00
    $134.00
    Pack of 200
       $33.50
    Case of 800
       $134.00
    Supplier Number: 72.706.600
  • 77588-932
    Low DNA-binding surface
    Low DNA-binding surface
    Transparent
    1.5 ml
    Transparent, PP
    Non sterile (PCR performance tested)
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 200
    Pack of 200 - $33.50
    $33.50
    Log in for availability
    Case of 800
    Case of 800 - $134.00
    $134.00
    Pack of 200
       $33.50
    Case of 800
       $134.00
    Supplier Number: 72.706.700
  • 77588-852
    Low protein-binding surface
    Low protein-binding surface
    Transparent
    2 ml
    Transparent, PP
    Non sterile (PCR performance tested)
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 200
    Pack of 200 - $36.78
    $36.78
    Log in for availability
    Case of 800
    Case of 800 - $147.12
    $147.12
    Pack of 200
       $36.78
    Case of 800
       $147.12
    Supplier Number: 72.695.600
  • 77588-854
    Low DNA-binding surface
    Low DNA-binding surface
    Transparent
    2 ml
    Transparent, PP
    Non sterile (PCR performance tested)
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 200
    Pack of 200 - $33.50
    $33.50
    Log in for availability
    Case of 800
    Case of 800 - $134.00
    $134.00
    Pack of 200
       $33.50
    Case of 800
       $134.00
    Supplier Number: 72.695.700