Cap with Volume Limitation
Catalog # 77588-150
Supplier: SARSTEDT INC
New Product
Specifications
- Material:PP
- Cat. No.:77588-150
- Cap color:Neutral
Specifications
About this item
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Designed for volume limitation to 100 µl
- Suitable for use with screw cap micro tube 72.733.201
- Polypropylene material
- Ribbed surface for maximum grip
Designed for the safe storage of even the smallest volumes, this cap with volume limitation ensures a maximum recovery rate of the sample material.