Orders LinkContactUsLinkComponent
Storage Boxes, PP
Storage Boxes, PP
Storage Boxes, PP
Storage Boxes, PP
Storage Boxes, PP
High-quality, safe, and robust racks and boxes are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Suitable for vessels with maximum dimensions of 45×12 mm
  • For storage down to –86 °C
  • Numerical coding makes it easy to identify samples
  • Various box designs for tubes up to max. Ø 12 mm
  • Can be autoclaved at 121 °C
  • Made of high-quality polypropylene
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application.
Storage boxes with a 9×9 configuration can accommodate 81 total items. Boxes with a 10×10 configuration can accommodate 100 total items. Assorted color options contain a mix of red, green, yellow, blue and orange.
Product Family Options
  • Description
    Material
  • Storage box with slip-on lid, 9×9 format
    Polypropylene
    Pack of 5
    Pack of 5 - $50.39
    $50.39
    Case of 20
    Case of 20 - $201.56
    $201.56
    Pack of 5
       $50.39
    Case of 20
       $201.56
     : 93.876.681
  • Storage box with slip-on lid, 9×9 format
    Polypropylene
    Pack of 5
    Pack of 5 - $50.39
    $50.39
    Case of 20
    Case of 20 - $201.56
    $201.56
    Pack of 5
       $50.39
    Case of 20
       $201.56
     : 93.876.581
  • Storage box with slip-on lid, 9×9 format
    Polypropylene
    Pack of 5
    Pack of 5 - $50.39
    $50.39
    Case of 20
    Case of 20 - $201.56
    $201.56
    Pack of 5
       $50.39
    Case of 20
       $201.56
     : 93.876
  • Storage box with slip-on lid, 9×9 format
    Polypropylene
    Pack of 5
    Pack of 5 - $50.39
    $50.39
    Case of 20
    Case of 20 - $201.56
    $201.56
    Pack of 5
       $50.39
    Case of 20
       $201.56
     : 93.876.381
  • Storage box with slip-on lid, 9×9 format
    Polypropylene
    Pack of 5
    Pack of 5 - $50.39
    $50.39
    Case of 20
    Case of 20 - $201.56
    $201.56
    Pack of 5
       $50.39
    Case of 20
       $201.56
     : 93.876.281
  • Storage box with slip-on lid, 9×9 format
    Polypropylene
    Pack of 5
    Pack of 5 - $50.39
    $50.39
    Case of 20
    Case of 20 - $201.56
    $201.56
    Pack of 5
       $50.39
    Case of 20
       $201.56
     : 93.876.481
  • Storage box with hinged lid, 10×10 format
    Polypropylene
    Pack of 5
    Pack of 5 - $52.22
    $52.22
    Case of 20
    Case of 20 - $208.88
    $208.88
    Pack of 5
       $52.22
    Case of 20
       $208.88
     : 93.877.610
  • Storage box with hinged lid, 10×10 format
    Polypropylene
    Pack of 5
    Pack of 5 - $52.22
    $52.22
    Case of 20
    Case of 20 - $208.88
    $208.88
    Pack of 5
       $52.22
    Case of 20
       $208.88
     : 93.877.510
  • Storage box with hinged lid, 10×10 format
    Polypropylene
    Pack of 5
    Pack of 5 - $52.22
    $52.22
    Case of 20
    Case of 20 - $208.88
    $208.88
    Pack of 5
       $52.22
    Case of 20
       $208.88
     : 93.877
  • Storage box with hinged lid, 10×10 format
    Polypropylene
    Pack of 5
    Pack of 5 - $52.22
    $52.22
    Case of 20
    Case of 20 - $208.88
    $208.88
    Pack of 5
       $52.22
    Case of 20
       $208.88
     : 93.877.310
  • Storage box with hinged lid, 10×10 format
    Polypropylene
    Pack of 5
    Pack of 5 - $52.22
    $52.22
    Case of 20
    Case of 20 - $208.88
    $208.88
    Pack of 5
       $52.22
    Case of 20
       $208.88
     : 93.877.210
  • Storage box with hinged lid, 10×10 format
    Polypropylene
    Pack of 5
    Pack of 5 - $52.22
    $52.22
    Case of 20
    Case of 20 - $208.88
    $208.88
    Pack of 5
       $52.22
    Case of 20
       $208.88
     : 93.877.410
  • Storage box with hinged lid, 10×10 format
    Polypropylene
    Pack of 5
    Pack of 5 - $53.59
    $53.59
    Case of 20
    Case of 20 - $214.36
    $214.36
    Pack of 5
       $53.59
    Case of 20
       $214.36
     : 93.877.999