Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Reaction Tubes

Reaction Tubes
 
Reaction Tubes
Reaction Tubes

 

 

High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.

  • Polypropylene retaining loop caps attached
  • Polypropylene tube material
  • Easy to open, secure seal when closed
  • Larger packaging units for economical storage and stocking
  • High centrifugation up to 20000×g

This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base; with or without skirted base; with push cap or screw cap; with or without print or labels.

Assorted color options contain a mix of red, green, blue, purple, yellow and neutral.

Product Family Options

 

  • Description
    Color
    Volume
     
     
  • Reaction tube
    Assorted
    1.5 ml
       
       
       
    Pack of 1,200
    Pack of 1,200 - $92.94
    $92.94
       
    Case of 6000
    Case of 6000 - $464.70
    $464.70
    Pack of 1,200
       $92.94

    Case of 6000
       $464.70

     : 72.690.999
  • Reaction tube
    Blue
    1.5 ml
       
       
       
    Pack of 500
    Pack of 500 - $32.81
    $32.81
       
    Case of 5000
    Case of 5000 - $328.10
    $328.10
    Pack of 500
       $32.81

    Case of 5000
       $328.10

     : 72.690.375
  • Reaction tube
    Brown
    1.5 ml
       
       
       
    Pack of 500
    Pack of 500 - $32.81
    $32.81
       
    Case of 5000
    Case of 5000 - $328.10
    $328.10
    Pack of 500
       $32.81

    Case of 5000
       $328.10

     : 72.690.304
  • Reaction tube
    Green
    1.5 ml
       
       
       
    Pack of 500
    Pack of 500 - $32.81
    $32.81
       
    Case of 5000
    Case of 5000 - $328.10
    $328.10
    Pack of 500
       $32.81

    Case of 5000
       $328.10

     : 72.690.376
  • Reaction tube
    Purple
    1.5 ml
       
       
       
    Pack of 500
    Pack of 500 - $32.81
    $32.81
       
    Case of 5000
    Case of 5000 - $328.10
    $328.10
    Pack of 500
       $32.81

    Case of 5000
       $328.10

     : 72.690.307
  • Reaction tube
    Red
    1.5 ml
       
       
       
    Pack of 500
    Pack of 500 - $32.81
    $32.81
       
    Case of 5000
    Case of 5000 - $328.10
    $328.10
    Pack of 500
       $32.81

    Case of 5000
       $328.10

     : 72.690.377
  • Reaction tube
    Transparent
    1.5 ml
       
       
       
    Pack of 500
    Pack of 500 - $32.81
    $32.81
       
    Case of 5000
    Case of 5000 - $328.10
    $328.10
    Pack of 500
       $32.81

    Case of 5000
       $328.10

     : 72.690.300
  • Reaction tube
    Transparent
    1.5 ml
       
       
       
    Pack of 500
    Pack of 500 - $32.81
    $32.81
       
    Case of 5000
    Case of 5000 - $328.10
    $328.10
    Pack of 500
       $32.81

    Case of 5000
       $328.10

     : 72.690.301
  • Reaction tube
    Yellow
    1.5 ml
       
       
       
    Pack of 500
    Pack of 500 - $32.81
    $32.81
       
    Case of 5000
    Case of 5000 - $328.10
    $328.10
    Pack of 500
       $32.81

    Case of 5000
       $328.10

     : 72.690.378