Membrane Screw Cap
Catalog # 77588-104
Supplier: SARSTEDT INC
New Product
Specifications
- Description:Membrane screw cap
- Material:PP
- Cat. No.:77588-104
- Cap color:Neutral
Specifications
About this item
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Polypropylene material
- Pierceable lid due to integrated membrane
- Ideal for automated filling and direct adaptation to analyzers
- Can be used on all SARSTEDT screw cap micro tubes
- Ribbed surface for maximum grip
The membrane screw cap is designed for automated filling of screw cap micro tubes and direct adaptation to analyzers.