Protective Cover for Block Rack D17
77589-726
New Product
- Color:Grey
- Description:Protective cover for block rack D17
- Material:ABS
- Cat. No.:77589-726
High-quality, safe, and robust racks and boxes are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
- Suitable for use with Block Rack D17
- ABS material
- Can be autoclaved at 121 °C
- For opened tubes during sample storage
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application.
This high-strength acrylonitrile butadiene styrene (ABS) block rack cover prevents opened tubes from becoming contaminated.