Terasaki Microtest Plate, 60-Well
77589-408
New Product
- Sterility:Sterile
- Color:Transparent
- Description:Terasaki microtest plate, 60-well, TC-treated
- Material:PS
- Cat. No.:77589-408
- Bottom style:Conical base
SARSTEDT offers microtest plates for a variety of applications.
- 60-well plate for analyses with minute specimen volumes (maximum 10 µl)
- Made of crystal-clear polystyrene
- Supplied with cover plate, assembled
SARSTEDT plates are ideal for sample processing on automated systems.