Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
CryoRack

CryoRack

CryoRack, 100×210×22 mm
Catalog # 77589-724
Supplier:   SARSTEDT INC
Supplier #:  93.856.040
CryoRack
Catalog # 77589-724
Supplier:  SARSTEDT INC
CryoRack
CryoRack

CryoRack, 100×210×22 mm

Price
$34.02
Log in for availability
Pack of 1 Case of 10
Product Details & Documents

High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.

  • Suitable for use with CryoPure tubes
  • Polypropylene work rack for a total of 40 CryoPure tubes (10×4 format)
  • Alphanumeric coding for easy sample identification

For the storage of cell materials and cell components at temperatures down to -196°C, SARSTEDT offers the CryoPure certified freezing system to prevent the exposure of samples to additional risks of contamination.

The CryoRack features non-slip rubber feet and a special base lock to allow one-handed use. Alphanumeric coding is included to facilitate sample identification.

Specifications
  • Description:
    CryoRack
  • Dimensions:
    100×210×22 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.