Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Reaction Tubes without Cap

Reaction Tubes without Cap

Supplier:   SARSTEDT INC
Reaction Tubes without Cap
Supplier:  SARSTEDT INC
Certificates
Reaction Tubes without Cap
Reaction Tubes without Cap

4 Options Available Below

Product Details & Documents

High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.

  • Polypropylene tubes supplied without closures
  • Particularly suitable for purification steps and aliquoting processes
  • Good visibility of the sample
  • Surface offers sufficient space for labelling

This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base; with or without skirted base; with push cap or screw cap; with or without print or labels.

Product Family Options

Product Information

  • Color
    Volume
    Availability
    Price
  • 77588-860
    Reaction tubes without cap
    Reaction tubes without cap
    Transparent
    0.5 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $40.40
    $40.40
    Log in for availability
    Case of 5000
    Case of 5000 - $404.00
    $404.00
    Pack of 500
       $40.40

    Case of 5000
       $404.00

    Supplier Number: 72.698.200
  • 77588-856
    Reaction tubes without cap
    Reaction tubes without cap
    Transparent
    1.5 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $31.51
    $31.51
    Log in for availability
    Case of 10000
    Case of 10000 - $630.20
    $630.20
    Pack of 500
       $31.51

    Case of 10000
       $630.20

    Supplier Number: 72.696
  • 77588-934
    Reaction tubes without cap
    Reaction tubes without cap
    Transparent
    2 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $53.64
    $53.64
    Log in for availability
    Case of 5000
    Case of 5000 - $536.40
    $536.40
    Pack of 500
       $53.64

    Case of 5000
       $536.40

    Supplier Number: 72.708
  • 77588-936
    Reaction tubes without cap, with skirted base
    Reaction tubes without cap, with skirted base
    Transparent
    2 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $53.64
    $53.64
    Log in for availability
    Case of 5000
    Case of 5000 - $536.40
    $536.40
    Pack of 500
       $53.64

    Case of 5000
       $536.40

    Supplier Number: 72.709
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.