Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Sulphite Standard Solution

Sulfite standard solution, pack of 16×10 ml voluette ampoules (NIST), 5000 mg/L as SO₃
Catalog # MSPP-2267410
Marketsource
Supplier:   Hach
 :  2267410
Sulphite Standard Solution
Catalog # MSPP-2267410
Supplier:  Hach
Marketsource
CAS Number:  
Sulphite Standard Solution
Sulphite Standard Solution

Sulfite standard solution, pack of 16×10 ml voluette ampoules (NIST), 5000 mg/L as SO₃

CAS Number:  
Price
$150.92
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

This equivalent standard is prepared with stabilized sodium thiosulfate, since sulphite is not a stable in solution.

Specifications

Specifications

  • Concentration:
    5000 mg/L as SO₃
  • Description:
    Sulfite standard solution, pack of 16×10 ml voluette ampoules (NIST)

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.