Alkalinity Standard Solution, 25000 mg/L CaCO₃ (NIST)
Catalog # MSPP-1427810
Supplier: HACH COMPANY MS
CAS Number:
Specifications
- Concentration:25000 mg/L as CaCO₃
- Pack type:10 ml Voluette ampoules
- Description:Alkalinity standard solution
- Cat. No.:MSPP-1427810
Specifications
About this item
Standard solution for accuracy checks in alkalinity determinations.
- Simple accuracy checks
- Minimize waste
- Reliable results
Voluette and PourRite ampule standards are designed to work directly with standard additions accuracy checks in Hach procedures and photometer software. Use the Tensette pipet to add precise spikes of standard and check percent recovery, to confirm analytical system performance.