Spleen Dissociation Medium
Catalog # MSPP-07915
Supplier: Stemcell Technologies
CAS Number:
Specifications
- Species:Mouse
- Description:Spleen dissociation medium
- Size:40 ml
- Cat. No.:MSPP-07915
Specifications
About this item
Medium for dissociation of mouse spleen.
- Maximize recovery and viability of dendritic cells from mouse spleen
- Optimized for use with selected EasySep™ cell separation kits
Maximize the recovery of dendritic cells from mouse spleens with Spleen dissociation medium, when used in combination with EasySep™ cell separation technology. This enzymatic dissociation medium contains collagenase, DNase, and fetal bovine serum (FBS), and has been optimized to maximize the viability of isolated spleen dendritic cells. Each 4 ml tube is sufficient for processing up to two mouse spleens.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').