Order Entry
Puerto Rico
ContactUsLinkComponent
Sephadex™ G-10 Gel Filtration Media, Cytiva
Sephadex™ G-10 Gel Filtration Media, Cytiva
Supplier:  Cytiva
Certificates
undefined
Sephadex™ G-10 Gel Filtration Media, Cytiva
Supplier:  Cytiva
Certificates

Specifications

  • pH Range:
    2 - 13
  • Matrix:
    Cross-linked dextran
  • Fractionation Range:
    <7 x 10² dextrans, <7 x 10² globular proteins
  • Storage Temperature:
    4 - 30 °C

Specifications