Order Entry
Puerto Rico
ContactUsLinkComponent
Gentle Cell Dissociation Reagent
Gentle Cell Dissociation Reagent
Catalog # MSPP-100-0485
CAS Number:  
undefined
Gentle Cell Dissociation Reagent
Catalog # MSPP-100-0485
Supplier Number:  100-0485
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Species:
    Human
    Mouse
  • Description:
    Gentle cell dissociation reagent
  • Size:
    100 ml
  • Cat. No.:
    MSPP-100-0485

Specifications

About this item

cGMP, enzyme-free cell dissociation reagent.

  • Obtain quality cultures with this gentle, chemically-defined, enzyme-free, GMP solution
  • Achieve high expansion of human ES/iPS cells during routine culture
  • Simplify cell passaging with straightforward, room temperature passaging protocols

Use Gentle Cell Dissociation Reagent (GCDR) to dissociate human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells into cell aggregates for routine passaging, or into a single-cell suspension. This enzyme-free and chemically-defined reagent ensures high expansion of human ES and iPS cells during routine culture. It is also suitable for the isolation of intestinal crypts to establish intestinal organoids, and for breaking up Corning® Matrigel® domes when passaging organoid cultures.

GCDR is now also manufactured following relevant cGMPs under a certified quality management system to ensure the highest quality and consistency for reproducible results.

Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').