- Species:HumanMouseNon-human primateRat
- Description:Collagenase B, ACF
- Size:1 g
- Cat. No.:MSPP-07439
Animal component-free collagenase for the digestion of native collagen fibrils.
- Obtain cleaner cultures with an animal component-free solution
Use Collagenase B, Animal Component-Free (ACF) to digest native collagen fibrils in connective tissues and prevent the introduction of animal-derived pathogens. This enzymatic preparation is sourced from Clostridium histolyticum and is obtained from cultures free of animal-derived materials. Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler and Yaron). Collagenase B, ACF contains higher collagenase and caseinase levels than Collagenase A.