Matrix for maintenance of human ES and iPS cells in combination with TeSR™ maintenance media.
- Decrease sources of variability in experiments with a recombinant human protein matrix
- Passage cells without the need for apoptotic inhibitors
- Use with any TeSR™ family medium to maintain hPSCs
- Increase single-cell attachment and survival when using with eTeSR™ for single-cell passaging
- Mimic the stem cell niche with this physiologically-relevant matrix
Support the growth and differentiation of human embryonic stem (ES) and induced pluripotent stem (iPS) cells under feeder-free conditions with this defined and xeno-free cell culture matrix.
For consistent cell populations and reproducible results in downstream applications, use CellAdhere™ Laminin-521 with TeSR™ maintenance media to provide a defined culture substrate for cell maintenance. Laminin 521 is expressed and secreted by human pluripotent stem cells (hPSCs) in the inner cell mass of the embryo and therefore creates a biologically relevant hPSC culture environment in vitro. Use CellAdhere™ Laminin-521 with eTeSR™ (MSPP-100-1215) maintenance medium for single-cell passaging. Compared to other matrices, CellAdhere™ Laminin-521 increases single-cell attachment and survival and does not require the addition of apoptotic inhibitors during long term culture. Pair with Gentle Cell Dissociation Reagent (GCDR;MSPP-07174) or ReLeSR™ (MSPP-05872) for routine passaging of PSC aggregates, or Accutase™ (MSPP-07920) for single-cell passaging workflows.
Note that single-cell passaging of human ES and iPS cells can result in selective pressure and lead to genetic aberrations. If passaging as single cells, we recommend checking the karyotype frequently.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Species:Human