Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Base/Neutral Surrogate Standards for CLP Series Methods
Base/Neutral Surrogate Standards for CLP Series Methods
Base/Neutral Surrogate Standards for CLP Series Methods
Catalog # SXCLP90-SB5
Supplier:
SPEX CERTIPREP LLC
New Product
CAS Number:
Expand image
Base/Neutral Surrogate Standards for CLP Series Methods
Catalog # SXCLP90-SB5
Supplier:
SPEX CERTIPREP LLC
Supplier Number:
CLP90-SB5
New Product
CAS Number:
Specifications
Size:
5 ml
Cat. No.:
SXCLP90-SB5
Storage temperature:
Ambient
Specifications
About this item
Product Details
Product Details