Order Entry
Puerto Rico
ContactUsLinkComponent
Krebs-Henseleit Buffer
Krebs-Henseleit Buffer
Catalog # 77442-756
Supplier:  Teknova
CAS Number:  
Krebs-Henseleit Buffer
Catalog # 77442-756
Supplier:  Teknova
Supplier Number:  K2352
CAS Number:  

Specifications

  • Size:
    1000 ml
  • Shelf Life:
    1095 days
  • Storage Temperature:
    Store at room temperature
  • Cat. No.:
    77442-756

Specifications

About this item

Krebs-Henseleit buffer composition: 0.200% Cerelose anhydrous dextrose (Glucose), 0.014% magnesium sulfate, 0.016% monopotassium phosphate, 0.035% potassium chloride, 0.690% sodium chloride, 373 µg/ml calcium chloride and 0.210% sodium carbonate.