Order Entry
Puerto Rico
ContactUsLinkComponent
D-(+)-Glucose solution 45% in aqueous solution, sterile
D-(+)-Glucose solution 45% in aqueous solution, sterile
Catalog # 77442-768
Supplier:  Teknova
CAS Number:  
D-(+)-Glucose solution 45% in aqueous solution, sterile
Catalog # 77442-768
Supplier:  Teknova
Supplier Number:  G4045
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Cat. No.:
    77442-768

Specifications

About this item

 :
Boiling Pt:100 °C
 :180.16 g/mol
Storage Temperature:Room Temperature
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912