Order Entry
Puerto Rico
ContactUsLinkComponent
pDNA Equilibration Buffer, pH 7.2
pDNA Equilibration Buffer, pH 7.2
Catalog # 77509-586
Supplier:  Teknova
CAS Number:  
pDNA Equilibration Buffer, pH 7.2
Catalog # 77509-586
Supplier:  Teknova
Supplier Number:  21231
CAS Number:  

Specifications

  • Size:
    500 ml
  • Shelf Life:
    1095 days
  • Storage Temperature:
    Store at room temperature
  • Cat. No.:
    77509-586

Specifications

About this item

pDNA Equilibration buffer: 50 mM Tris, 10 mM EDTA.