Order Entry
Puerto Rico
ContactUsLinkComponent
pDNA Resuspension Buffer, pH 8.0
pDNA Resuspension Buffer, pH 8.0
Catalog # 77509-556
Supplier:  Teknova
CAS Number:  
pDNA Resuspension Buffer, pH 8.0
Catalog # 77509-556
Supplier:  Teknova
Supplier Number:  21216
CAS Number:  

Specifications

  • Size:
    1000 ml
  • Shelf Life:
    1095 days
  • Storage Temperature:
    Store at room temperature
  • Cat. No.:
    77509-556

Specifications

About this item

pDNA Resuspension buffer composition: 50 mM Tris/Tris-HCl, 10 mM EDTA.