HNE Buffer, Sterile
1000 ml, 1 Bottle per pack
Catalog # 77623-564
Supplier: Teknova
Supplier #: 21509
Jump to:
Product Details & Documents
HNE Buffer composition: 50 mM HEPES, 0.15 M sodium chloride, 25 mM EDTA.
Specifications
Specifications
- Pack type:1 Bottle per pack
- Size:1000 ml
- Cat. No.:77623-564
- Shelf Life:1095 days
- Storage Temperature:Store at room temperature