Pierce™ Fish Serum Blocking Buffe, Thermo Scientific
Catalog # PI37527
Supplier: Invitrogen
CAS Number:
Specifications
- Description:Fish Serum Blocking Buffer in Buffered Saline and 0.1% Sodium Azide
- Size:500 mL
- Environmentally Preferable:
- Cat. No.:PI37527
- Application:ELISA, Western Blot, Dot Blot
- Supplier no.:37527
Specifications
About this item
This Fish Serum Blocking Buffer is Steelhead Salmon Serum in PBS that is especially useful as a blocking agent in IHC and other detection methods involving mammalian samples.
- Non mammalian
- Easy to use
- Convenient, filtered and stabilized in PBS
Fish Serum Blocking Buffer is effective in stabilizing solutions and samples for antibody-binding interactions while at the same time minimizing the possibility of cross-reaction with mammalian sample components
The blocker has been used to decrease background and increase signal-to-noise ratios with near IR fluorescent probes and is validated for use with the LI-COR Odyssey™ Infrared Imaging System.
The product is also especially effective in various cellular imaging protocols based on visible and near infrared fluorescence detection.