Orders LinkContactUsLinkComponent
Vitrectomy Scissors
Vitrectomy Scissors
Vitrectomy Scissors
Certificates
Vitrectomy Scissors
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Vitrectomy scissors are pencil-style scissors with a new, ergonomic handle design with micro-blades that are easy to clean. Two models available, straight and angled. These scissors designed for vitrectomy, an ophthalmalogical procedure.
Vitrectomy is the surgical removal of some or all of the vitreous humor, the gel that fills the eyeball, in order to correct retinal issues like retinal tears, macular holes, floaters and more. During surgery, an ophthalmologist penetrates the white of the eye, the sclera, and inserts the instruments necessary to complete the surgery.
Product Family Options
Product Information
  • Material
    Blade type
    Availability
    Price
  • 77665-746
    Vitrectomy scissors, 20 G
    Vitrectomy scissors, 20 G
    Stainless steel
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $448.00
    $448.00
    Each
       $448.00
    Supplier Number: WP405200
  • 77665-748
    Vitrectomy scissors, 35°
    Vitrectomy scissors, 35°
    Stainless steel
    Angled
    Log in for availability
    Log in for availability
    Each
    Each - $327.00
    $327.00
    Each
       $327.00
    Supplier Number: WP405240
  • 77666-308
    Vitrectomy scissors, 65°, 20 G
    Vitrectomy scissors, 65°, 20 G
    Stainless steel
    Angled
    Log in for availability
    Log in for availability
    Each
    Each - $448.00
    $448.00
    Each
       $448.00
    Supplier Number: WP504240