Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium nitrate
Potassium nitrate ≥99.99% (trace metals basis), SPEX Infinitas™
Potassium nitrate ≥99.99% (trace metals basis), SPEX Infinitas™
:
SPEX CERTIPREP LLC
New Product
Potassium nitrate ≥99.99% (trace metals basis), SPEX Infinitas™
:
SPEX CERTIPREP LLC
New Product
:
7757-79-1
:
:
KNO₃
:
MFCD00011409
:
101.1 g/mol
:
400 °C (752 °F)
:
334 °C (633.2 °F)
:
2.109 g/cm³ (17.59961 lbs./gal)
:
1486
:
5.1,III
:
GHS03