Regular Unleaded Gas-87 Octane
Catalog # SXGAS-20
Supplier: SPEX CERTIPREP LLC
New Product
CAS Number:
Specifications
- Concentration:20 mg/ml
- Matrix:Methanol-P and T
- Size:1 ml
- Storage Temperature:Ambient
- Cat. No.:SXGAS-20
Specifications
About this item
GAS-20 is a standard of Octane 87 gasoline for use in monitoring gasoline intrusion or pollution in the environment including water, soil and other environmental matrices.