Volatiles Mix D for CLP SOW 2/88 and 3/90 (High Level)
Catalog # SXCLPV-DH
Supplier: SPEX CERTIPREP LLC
New Product
CAS Number:
Specifications
- Concentration:2000 µg/ml
- Matrix:Methanol-P and T
- Storage Temperature:Freezer
- Cat. No.:SXCLPV-DH
- Volume:1 ml
Specifications
About this item
Volatiles, CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.