Order Entry
Puerto Rico
ContactUsLinkComponent
High Concentration Acid Surrogates for CLP Series Methods
High Concentration Acid Surrogates for CLP Series Methods
Catalog # SXCLP90-75SA5
Supplier:  SPEX CERTIPREP LLC
New Product
CAS Number:  
undefined
High Concentration Acid Surrogates for CLP Series Methods
Catalog # SXCLP90-75SA5
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CLP90-75SA5
New Product
CAS Number:  

Specifications

  • Concentration:
    7500 µg/ml
  • Matrix:
    Methanol
  • Storage Temperature:
    Freezer
  • Cat. No.:
    SXCLP90-75SA5
  • Volume:
    5 ml

Specifications

About this item

CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.