Polynuclear Aromatic Hydrocarbon (PAH) Combination Mix
Catalog # SX8310-A
Supplier: SPEX CERTIPREP LLC
New Product
CAS Number:
Specifications
- Cat. No.:SX8310-A
- Storage temperature:Ambient
Specifications
About this item
US EPA method 8310 is an analytical method for the monitoring of PNA hydrocarbons in groundwater and waste samples by HPLC using UV and fluorescence detectors.