Order Entry
Puerto Rico
ContactUsLinkComponent
Long List Appendix IX Compound Mix E (High Level)
Long List Appendix IX Compound Mix E (High Level)
Catalog # SX8260-EH
Supplier:  SPEX CERTIPREP LLC
New Product
CAS Number:  
undefined
Long List Appendix IX Compound Mix E (High Level)
Catalog # SX8260-EH
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  8260-EH
New Product
CAS Number:  

Specifications

  • Cat. No.:
    SX8260-EH
  • Storage temperature:
    Freezer

Specifications

About this item

US EPA method 8260 is a general analytical method for the monitoring of VOCs by GC/MS. CLP series methods - The US EPA retains analytical services through the contract laboratory program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP volatile organic analysis method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.