Phthalates Esters Semivolatiles Mix
Catalog # SX8061-X
Supplier: SPEX CERTIPREP LLC
New Product
CAS Number:
Specifications
- Cat. No.:SX8061-X
- Storage temperature:Ambient
Specifications
About this item
US EPA method 8061 is an analytical method for the monitoring of phthalate esters in aqueous and solid samples by GC/MS or GC/ECD.