Order Entry
Puerto Rico
ContactUsLinkComponent
 
Alcohols Mix
Catalog # SX8015B-A
Supplier:  SPEX CERTIPREP LLC
New Product
CAS Number:  
undefined
Alcohols Mix
Catalog # SX8015B-A
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  8015B-A
New Product
CAS Number:  

Specifications

  • Cat. No.:
    SX8015B-A
  • Storage temperature:
    Refrigerator

Specifications

About this item

US EPA method 8015 is a general analytical method for the monitoring of certain non-halogenated VOCs and SVOCs by GC/FID.