Order Entry
Puerto Rico
ContactUsLinkComponent
Method 524.2 Revision 4 Mix
Method 524.2 Revision 4 Mix
Catalog # SX5242-R4
Supplier:  SPEX CERTIPREP LLC
New Product
CAS Number:  
undefined
Method 524.2 Revision 4 Mix
Catalog # SX5242-R4
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  5242-R4
New Product
CAS Number:  

Specifications

  • Cat. No.:
    SX5242-R4
  • Storage temperature:
    Freezer

Specifications

About this item

US EPA method 524.3 is a GC/MS method for the analysis of purgeable VOCs in finished drinking water. Analytes added to methanol 524.3 revision 4 are supplied in a 24-component mix.