Acrolein and Acrylonitrile Standards
SX603-XM
New Product
:
- Cat. No.:SX603-XM
- Storage temperature:Freezer
US EPA method 603 is a general analytical method for the separation of acrolein and acrylonitrile in municipal and industrial wastewater by GC/FID.