Bulwark® Women's FR/CP Lab Coats
Supplier: Bulwark Protection
New Product
Certificates
About this item
This FR/CP lab coat brings together the latest innovations in lab safety to protect wearers against multiple hazards. Flame resistant fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame. FR/CP garments also repel small amounts of liquid chemicals that are either dropped or splashed on the fabric, resisting penetration and wicking of an inadvertent, secondary liquid chemical splash. It is also lightweight, breathable and comfortable to wear.
- Hazard
- Wash care: Industrial laundry - light soil
- Fabric: Body: 4.5 oz. flame resistant, chemical protection
- Blend: Body: 93% Meta-aramid/5% para-aramid/2% anti-stat
- Concealed snap closure
- Black lay down collar for easy recognition as a CP lab coat
- FR-only knit cuffs
- Gender: Female
Splash resistant pass-throughs for easy inner pocket access. Pockets include one two-needle top-stitched chest pocket with pencil stall and two double-needle top-stitched lower pockets with flaps.
Certifications: Certifications level 1: NFPA 2112. Certifications level 2: Category 1 protection.
Specifications
- Catalog No:
- 77661-310
- 77661-312
- 77661-314
- 77661-318
- 77661-308
- 77661-316
- Fabric:
- Body: 4.5 oz. Flame resistant, chemical protection
- Body: 4.5 oz. Flame resistant, chemical protection
- Body: 4.5 oz. Flame resistant, chemical protection
- Body: 4.5 oz. Flame resistant, chemical protection
- Body: 4.5 oz. Flame resistant, chemical protection
- Body: 4.5 oz. Flame resistant, chemical protection
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long