ContactUsLinkComponent
Erlenmeyer Flasks, PP
Erlenmeyer Flasks, PP
  77674-458
 :  Brandtech
New Product
Erlenmeyer Flasks, PP
  77674-458
 :  Brandtech
 :  570941
New Product
  • Graduation Interval:
    200 ml
  • Capacity:
    1000 ml
  • Environmentally Preferable:
  • Cat. No.:
    77674-458
  • Thread:
    GL 45
Highly transparent well suited for storage and cultivation of cell cultures.
  • Suitable for microwaves
  • To preserve markings, cleaning at no higher than 60 °C is recommended
Far safer than glass flasks for use in incubator shakers due to the break resistance of plastic. Suitable for contact with foodstuffs according to regulation (EU) No. 10/2011.