Order Entry
Puerto Rico
ContactUsLinkComponent
Pierce™ Dye Removal Columns, Thermo Scientific
Pierce™ Dye Removal Columns, Thermo Scientific
Catalog # PI22858
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Dye Removal Columns, Thermo Scientific
Catalog # PI22858
Supplier:  Invitrogen
Supplier Number:  22858
CAS Number:  

Specifications

  • Description:
    Dye Removal Columns
  • Cat. No.:
    PI22858
  • Supplier no.:
    22858

Specifications

About this item

Pierce™ Dye Removal Columns enable fast and efficient removal of non-reacted fluorescent dyes from protein labeling reactions.

  • Removes free, unconjugated dye from labeled protein solutions in less than a minute
  • Purification resin provides outstanding conjugate recovery
  • Separate resin and microcentrifuge columns allow optimization for different dyes and concentrations of sample

Removing excess dye after a labeling reaction is often difficult and time-consuming but is essential for accurate determination of dye-to-protein ratios. The dye removal resin in this kit is highly specialized to produce exceptional protein recoveries while effectively removing non-conjugated dye. Using the appropriate amount of resin and buffer conditions, almost any fluorescent dye can be removed with this kit.