ContactUsLinkComponent
HALO® C18 OLIGO HPLC Columns
HALO® C18 OLIGO HPLC Columns
New Product
Certificates
HALO® C18 OLIGO HPLC Columns
New Product
Specifications
  • Catalog No:
  • 77660-608
  • 77660-610
  • 77660-612
  • 77660-614
  • 77660-616
  • 77660-618
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • pH Range:
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • Temperature Limit:
  • 90 °C (pH 2); 85 °C (pH 9)
  • 90 °C (pH 2); 85 °C (pH 9)
  • 90 °C (pH 2); 85 °C (pH 9)
  • 90 °C (pH 2); 85 °C (pH 9)
  • 90 °C (pH 2); 85 °C (pH 9)
  • 90 °C (pH 2); 85 °C (pH 9)
  • USP Code:
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1