VWR® Multi-Channel Polypropylene Reagent Reservoirs
Supplier: VWR International
New Product
Certificates
About this item
These Polypropylene multi-channel reagent reservoirs are designed to support both automated liquid handling platforms and manual systems. The reservoir design complies with ANSI/SLAS 1-2004, when used for cell culture and immunoassay experiments.
- Multiple capacities and well numbers can be selected for different experimental needs
- Multi-channel reagent reservoirs are suitable for both 8-channel and 12-channel pipettes
- Rhombic well series: 96- or 384-well reagent reservoirs on the bottom
- Non-sterile or sterilized by irradiation, SAL 10⁻⁶ (ISO 11137)
- Lot-certified, non-pyrogenic, DNase- and RNase-free
Product dimensions conform to SBS standards, and are highly adaptable and compatible with most automation systems.
Certifications: ISO 13485 and ISO 9001.
Specifications
- Catalog No:
- 77575-290
- 77575-302
- 77575-288
- 77575-300
- 77575-298
- 77575-296
- 77575-294
- 77575-292
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M