ContactUsLinkComponent
Logic® Cell Culture Cart Side Shelf Accessory
Logic® Cell Culture Cart Side Shelf Accessory
Catalog # 77574-614
Supplier:  Labconco
New Product
undefined
Logic® Cell Culture Cart Side Shelf Accessory
Catalog # 77574-614
Supplier:  Labconco
Supplier Number:  8024700
New Product
Specifications
  • Description:
    Logic cell culture cart side shelf accessory provides two accessory side shelves for extra storage
  • Cat. No.:
    77574-614
Specifications
About this item
Logic cell culture cart side shelf accessory provides two accessory side shelves for extra storage.
  • Extra storage for bottles, gloves and other required items
  • Can mount on either side of cart
  • Potential for four shelves total
These shelves provide extra storage for bottles, gloves and other required items. The can be mounted to the right or left side of the cart for up to four shelves total. Shipping weight 5 lbs.