Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ mouse PE positive selection kit II
- Cat. No.:MSPP-17666
Specifications
About this item
Immunomagnetic positive selection of mouse cells labeled with PE-conjugated antibodies.
- Fast and easy-to-use
- No columns required
The EasySep™ Mouse PE Positive Selection Kit II is designed to isolate cells that are labeled with PE-conjugated antibodies. Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This kit replaces the EasySep™ Mouse PE Positive Selection Kit.