EasySep™ Human TCR Alpha/Beta Depletion Kit
Catalog # MSPP-17847
Supplier: Stemcell Technologies
Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ human TCR alpha/beta depletion Kit
- Cat. No.:MSPP-17847
Specifications
About this item
Immunomagnetic depletion of human TCRa/b+ cells.
- Fast, easy-to-use, and column-free procedure
- Untouched, viable cells
The EasySep™ Human TCR Alpha/Beta Depletion Kit is designed to deplete human T cell receptor alpha/beta+ (TCRab+) cells from leukapheresis samples or expanded TCRa/b knockout cell cultures. TCRab+ cells are targeted with antibody complexes and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Labeled cells remain in the tube while unlabeled cells are poured off.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').