Specifications
- Capacity:20 ml biofluid processed
- Description:EasySep™ human pan-extracellular vesicle positive selection kit
- Cat. No.:MSPP-17891
Specifications
About this item
Fast and easy isolation of human extracellular vesicles using immunomagnetic positive selection.
- Fast, easy to use, and column-free
- Avoid the need for ultracentrifugation and associated time consuming EV isolation methods
- Obtain highly purified EVs without contaminating biofluid components
The EasySep™ Human Pan-Extracellular Vesicle Positive Selection Kit isolates highly purified human extracellular vesicles from plasma/serum and cell culture conditioned medium by immunomagnetic positive selection. Extracellular vesicles are labeled with Tetrameric Antibody Complexes recognizing CD9, CD81, and CD63, and magnetic particles. Labeled extracellular vesicles are separated without columns using an EasySep™ magnet. Unwanted biofluid components are simply poured off, while desired extracellular vesicles remain in the tube. Isolated extracellular vesicles are available for downstream applications such as DNA/RNA extraction, Western blot, or mass spectrometry.