ContactUsLinkComponent
EasySep™ Human Myeloid Positive Selection Kit II
EasySep™ Human Myeloid Positive Selection Kit II
Catalog # MSPP-17893
undefined
EasySep™ Human Myeloid Positive Selection Kit II
Catalog # MSPP-17893
Supplier Number:  17893
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ human myeloid positive selection kit II
  • Cat. No.:
    MSPP-17893
Specifications
About this item
Immunomagnetic positive selection of human CD33+ and CD66+ cells (e.g. myeloid cells).
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ Human Myeloid Positive Selection Kit II is designed to isolate CD33+ and CD66b+ cells from ammonium chloride lysed blood or bone marrow by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD33, CD66b and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD33 and CD66b antigens are expressed on myeloid progenitors monocytes and granulocytes.
This product replaces the EasySep™ Human Myeloid Positive Selection Kit for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').