Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ human neutrophil isolation kit
- Cat. No.:MSPP-17957
Specifications
About this item
Immunomagnetic isolation of untouched human neutrophils.
- Fast, easy-to-use and column-free
- Up to 99% purity
- Untouched, viable cells
The EasySep™ Human Neutrophil Isolation Kit is designed to isolate neutrophils from a polymorphonuclear cell-rich fraction of peripheral blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-neutrophils and magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. The isolation kit is compatible with cells prepared using HetaSep™ sedimentation or red blood cell lysis.
This product replaces the EasySep™ Human Neutrophil Enrichment Kit, for even faster cell isolations.